Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Oxygen and moisture removal column, for reduceing the oxygen level to less than 5 ppm and moisture level to less than 50 ppm; requires connection to two manual valves on the main chamber and a diaphragm vacuum pump for atmosphere oxygen/moisture removal and recirculation
Catalog # LBCP5244200
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5244200
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Oxygen and moisture removal column, for reduceing the oxygen level to less than 5 ppm and moisture level to less than 50 ppm; requires connection to two manual valves on the main chamber and a diaphragm vacuum pump for atmosphere oxygen/moisture removal and recirculation
Price
SFr. 3’460.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Oxygen and moisture removal column, for reduceing the oxygen level to less than 5 ppm and moisture level to less than 50 ppm; requires connection to two manual valves on the main chamber and a diaphragm vacuum pump for atmosphere oxygen/moisture removal and recirculation
  • Seq:
    20