Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Flat work surface, black epoxy resin, may be placed atop two Protector Standard Base Cabinets to support Precise™ glove boxes, 1524×762×31,75 mm
Catalog # LBCP4882807
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP4882807
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Flat work surface, black epoxy resin, may be placed atop two Protector Standard Base Cabinets to support Precise™ glove boxes, 1524×762×31,75 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Flat work surface, black epoxy resin, may be placed atop two Protector Standard Base Cabinets to support Precise™ glove boxes, 1524×762×31,75 mm
  • Seq:
    31
  • Cat. No.:
    LBCP4882807