Accessories for glove boxes, Precise™
Left side shelf kit, incl. four stainless steel shelves and epoxy-coated steel vertical uprights, for storage in left side of the main chamber to maximise space; assembled dimensions: 538,48×508×111,76 mm
Catalog # LBCP5235001
Supplier: LABCONCO
Jump to:
Product Details & Documents
Specifications
- Description:Left side shelf kit, incl. four stainless steel shelves and epoxy-coated steel vertical uprights, for storage in left side of the main chamber to maximise space; assembled dimensions: 538,48×508×111,76 mm
- Seq:44