ContactUsLinkComponent
SUB Aqua 26 Plus, 26 litre Digital unstirred water bath -Electronic PID controlled. Temp range: Ambient+5C to 99C. Temp stability at 37C: 0.2C. Mains supply: 230V AC 50/60Hz. Includes ploycarbonate lid, base tray and UK cordset 1 * 1 items
Catalog # 462-0193
Supplier:  GRANT INSTRUMENTS
SUB Aqua 26 Plus, 26 litre Digital unstirred water bath -Electronic PID controlled. Temp range: Ambient+5C to 99C. Temp stability at 37C: 0.2C. Mains supply: 230V AC 50/60Hz. Includes ploycarbonate lid, base tray and UK cordset 1 * 1 items
Catalog # 462-0193
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together