ContactUsLinkComponent
Magnetic stirrer, warming- and maintenance-free with 1 stirring point, large surface, for careful and protective mixing of cell susp ensions and culture broths, also suitable for viscous culture media. 1 * 1 items
Catalog # TMAG80001_CH
Supplier:  2MAG
Magnetic stirrer, warming- and maintenance-free with 1 stirring point, large surface, for careful and protective mixing of cell susp ensions and culture broths, also suitable for viscous culture media. 1 * 1 items
Catalog # TMAG80001_CH
Supplier:  2MAG
Frequently Bought Together