Caps
Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter, GL 45, 1 port for 1/16" tubing, 3 ports for 1/8" and 1/16" OD tubing, PP
Catalog # JOURJR-S-11003
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
These caps top bottles are available with two, three, or four colour-coded 1/4-28 thread for ¹/₈ or ¹/₁₆" O.Ø tubing.
Specifications
- Thread:GL 45
- Description:Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter
- Material:PP
- Connector:1 port for 1/16" tubing, 3 ports for 1/8" and 1/16" OD tubing