Caps
Cap kit, includes one cap, with fittings, one 2 l safety coated glass bottle, 3 m PFA tubing, one No-Met 5 µm mobile phase filter, GL 45, 1 port for 1/16" tubing, 2 ports for 1/8" and 1/16" OD tubing, PP
Catalog # JOURJR-S-11202
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
These caps top bottles are available with two, three, or four colour-coded 1/4-28 thread for ¹/₈ or ¹/₁₆" O.Ø tubing.
Specifications
- Thread:GL 45
- Description:Cap kit, includes one cap, with fittings, one 2 l safety coated glass bottle, 3 m PFA tubing, one No-Met 5 µm mobile phase filter
- Material:PP
- Connector:1 port for 1/16" tubing, 2 ports for 1/8" and 1/16" OD tubing