Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Stirring rods

Stirring rods, glass, with fused ends, Glass, 6 mm, 200 mm
  441-0457
Stirring rods
  441-0457
Stirring rods
Stirring rods

Stirring rods, glass, with fused ends, Glass, 6 mm, 200 mm

 
SFr. 30.40
   
1 * 50 ST
 

AR-Glas®. With fused ends.

 
  • Ø:
    6 mm
  • Description:
    Stirring rods, glass, with fused ends
  • Length:
    200 mm
  • Material:
    Glass
  • Seq:
    017