ContactUsLinkComponent
Holder thermo-conductive f 384-well PCR plate. CoolRack maintains temperature through direct contact with ice, dry ice, water, LN2 or water bath. Can be used with ThermalTray or CoolTray for extended temperature control. 1 * 1 items
Catalog # 479-0876
Supplier:  BIOCISION
Holder thermo-conductive f 384-well PCR plate. CoolRack maintains temperature through direct contact with ice, dry ice, water, LN2 or water bath. Can be used with ThermalTray or CoolTray for extended temperature control. 1 * 1 items
Catalog # 479-0876
Supplier:  BIOCISION
Frequently Bought Together