Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Test tubes without rim

Test tubes without rim

135 mm, 14,5 mm, 1 mm
Catalog # HECH42775042
Test tubes without rim
Catalog # HECH42775042
Test tubes without rim
Test tubes without rim

135 mm, 14,5 mm, 1 mm

Price
SFr. 31.50
Log in for availability
1 * 100 ST
Product Details & Documents

AR-Glas® (soda-lime glass).

  • Heavy wall (212-1700 suitable as Durham-Tube)
  • Good chemical and physical properties
  • Can also be used as disposable test tubes

Documents

Specifications
  • Ext.Ø:
    14,5 mm
  • Wall thickness:
    1 mm
  • Length:
    135 mm
  • Seq:
    16