ContactUsLinkComponent
Container, blue PS, 30mL, universal + spoon Screw cap PP with integral spoon CE marked 98/79/EC For use in vitro only Printed label Aseptically manufactured under cleanroom conditions Virgin PS, non-cytotoxic. 1 * 400 items
Catalog # SLIN128SBB
Supplier:  STERILIN
Container, blue PS, 30mL, universal + spoon Screw cap PP with integral spoon CE marked 98/79/EC For use in vitro only Printed label Aseptically manufactured under cleanroom conditions Virgin PS, non-cytotoxic. 1 * 400 items
Catalog # SLIN128SBB
Supplier:  STERILIN
Frequently Bought Together