ContactUsLinkComponent
Pcr tube, 0.2ml, thin wall, green, domed cap, nonsterile, autoclavable, virgin polypropylene, dnase, rnase, and pyrogen-free 1 * 1.000 items
Catalog # BITX3425.G.X
Anbieter:  BIOTIX
Pcr tube, 0.2ml, thin wall, green, domed cap, nonsterile, autoclavable, virgin polypropylene, dnase, rnase, and pyrogen-free 1 * 1.000 items
Catalog # BITX3425.G.X
Anbieter:  BIOTIX
Frequently Bought Together