Dip slides
Enterobact/Strepto + Neutr, 120
Catalog # LIOF53531
Supplier: LIOFILCHEM
Jump to:
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
- ISO 18593
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:120
- Description:Enterobact/Strepto + Neutr