Orders LinkContactUsLinkComponent
Dip slides
Dip slides
Total/Coliform + Neutr, 120
Catalog # LIOF53562
Supplier:   LIOFILCHEM
Dip slides
Catalog # LIOF53562
Supplier:  LIOFILCHEM
CAS Number:  
Dip slides
Dip slides
Total/Coliform + Neutr, 120
CAS Number:  
Price
SFr. 410.00
Log in for availability
1 * 120 ST
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
  • ISO 18593
Documents
Specifications
Specifications
  • Pk:
    120
  • Description:
    Total/Coliform + Neutr
Specifications