Pipette tips
Eppendorf type, 5 - 200 µl, +, Yellow, Eppendorf, Gilson, Socorex, Nichiryo, Biohit, Brand, Individually wrapped
: DELTALAB
- Packed:Individually wrapped
- Sterile:+
- Volume:5 - 200 µl
- Seq:10
- Tip style:Eppendorf type
- Tip colour:Yellow
- For use with:Eppendorf, Gilson, Socorex, Nichiryo, Biohit, Brand