Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Solid phase extraction cartridges, HyperSep™ Florisil

Cartridge, 40 - 60 µm, 15 ml, 2000 mg
Catalog # 549-0711
Solid phase extraction cartridges, HyperSep™ Florisil
Catalog # 549-0711
Solid phase extraction cartridges, HyperSep™ Florisil
Solid phase extraction cartridges, HyperSep™ Florisil

Cartridge, 40 - 60 µm, 15 ml, 2000 mg

Price
SFr. 241.00
Log in for availability
1 * 20 ST
Product Details & Documents

Ideal for the isolation of polar compounds from non polar matrices.

  • Magnesia-loaded silica gel offers rapid flow rates for large sample volumes
  • Extremely polar in nature, especially suited to the separation of chlorinated pesticides
Specifications
  • Column Format:
    Cartridge
  • Particle Size:
    40 - 60 µm
  • Capacity:
    15 ml
  • Seq:
    07
  • Bed weight:
    2000 mg