Orders LinkContactUsLinkComponent
Sample cups for analysers
Sample cups for analysers
Sample cups for auto sampler, PS, recommended for use with most aqueous solutions, 1,5 - 2 ml, Clear
Catalog # PERCB0119079
Supplier:   PerkinElmer
Sample cups for analysers
Catalog # PERCB0119079
Supplier:  PerkinElmer
Sample cups for analysers
Sample cups for auto sampler, PS, recommended for use with most aqueous solutions, 1,5 - 2 ml, Clear
Price
328.00 SFr.
Log in for availability
1 * 1.000 ST
Product Details & Documents
Disposable.
These sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample. For concentrated acids including HF for analyses.
Specifications
  • Capacity:
    1,5 - 2 ml
  • Colour:
    Clear
  • Description:
    Sample cups for auto sampler, PS, recommended for use with most aqueous solutions