Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Circular spacers

Circular Teflon spacers, 0,5 mm, FTIR, 1600 series
  PERC51001141
Circular spacers
  PERC51001141
Circular spacers

Circular Teflon spacers, 0,5 mm, FTIR, 1600 series

 
SFr. 242.00
   
1 * 3 ST
 

These circular spacers are designed to be used for the analysis of less viscous samples.

  • Available in 0,025 to 0,500 mm thickness

Since the demountable cells uses the circular windows, circular spacers should be used.

 
  • For:
    FTIR, 1600 series
  • Description:
    Circular Teflon spacers, 0,5 mm
  • Seq:
    04