Circular spacers
Circular Teflon spacers, 0,2 mm, FTIR, 1600 series
PERC51001140
These circular spacers are designed to be used for the analysis of less viscous samples.
- Available in 0,025 to 0,500 mm thickness
Since the demountable cells uses the circular windows, circular spacers should be used.
- For:FTIR, 1600 series
- Description:Circular Teflon spacers, 0,2 mm
- Seq:03



