Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Circular spacers

Circular spacers

Circular Teflon spacers, 0,05 mm, FTIR, 1600 series
Catalog # PERC51001135
Supplier:   PerkinElmer
Circular spacers
Catalog # PERC51001135
Supplier:  PerkinElmer
Circular spacers

Circular Teflon spacers, 0,05 mm, FTIR, 1600 series

Price
SFr. 210.00
Log in for availability
1 * 5 ST
Product Details & Documents

These circular spacers are designed to be used for the analysis of less viscous samples.

  • Available in 0,025 to 0,500 mm thickness

Since the demountable cells uses the circular windows, circular spacers should be used.

Specifications
  • For:
    FTIR, 1600 series
  • Description:
    Circular Teflon spacers, 0,05 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.