Orders LinkContactUsLinkComponent
LC-MS nitrogen generators
LC-MS nitrogen generators
N2-35 Mid Flow, without air compressor, 20 l/min at 100 psi; 25 l/min at 125 psi; 30 l/min at 100 psi; 37 l/min at 125 psi; 46 l/min at 100 psi; 57 l/min at 125 psi, 450×410×1300 mm
Catalog # PERCN9306211
Supplier:   PerkinElmer
LC-MS nitrogen generators
Catalog # PERCN9306211
Supplier:  PerkinElmer
LC-MS nitrogen generators
N2-35 Mid Flow, without air compressor, 20 l/min at 100 psi; 25 l/min at 125 psi; 30 l/min at 100 psi; 37 l/min at 125 psi; 46 l/min at 100 psi; 57 l/min at 125 psi, 450×410×1300 mm
Price
SFr. 53’830.00
Log in for availability
1 * 1 ST
Product Details & Documents
Nitrogen generators are designed to supply single or multiple LC-MS instruments with dry nitrogen at purities of 98 – 99,5%. The generator can also be used for solvent evaporation as well as supplying dry nitrogen to analytical instruments.
Specifications
  • Flow rate:
    20 l/min at 100 psi; 25 l/min at 125 psi; 30 l/min at 100 psi; 37 l/min at 125 psi; 46 l/min at 100 psi; 57 l/min at 125 psi
  • W×D×H:
    450×410×1300 mm
  • Type:
    N2-35 Mid Flow, without air compressor