Orders LinkContactUsLinkComponent
Closures for headspace vials
Closures for headspace vials
Red PTFE coated butyl cap/septa/spring
Catalog # PERCN9303991
Supplier:   PerkinElmer
Closures for headspace vials
Catalog # PERCN9303991
Supplier:  PerkinElmer
Closures for headspace vials
Red PTFE coated butyl cap/septa/spring
Price
SFr. 344.00
Log in for availability
1 * 1 KIT
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Specifications
  • Description:
    Red PTFE coated butyl cap/septa/spring
  • Seq:
    25