Sample cups for analysers
Sample cups for auto sampler, PP, recommended for blank solutions, reagent solutions and matrix modifiers, 7 ml, Translucent
Catalog # PERCB3001567
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Disposable.
These sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample. For concentrated acids including HF for analyses.
Specifications
- Capacity:7 ml
- Colour:Translucent
- Description:Sample cups for auto sampler, PP, recommended for blank solutions, reagent solutions and matrix modifiers
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..