Multi-parameter meter (pH/conductivity/oxygen), handheld, SensoDirect 150
Product Details & Documents
The SensoDirect 150 is designed for multi purpose operation and measures pH/Redox, dissolved oxygen and conductivity/TDS.
- Data logger function, results can be downloaded to a PC for further analysis
- Automatic calibration + buffer recognition
- Large LCD display, intuitive user interface, HOLD function
- Flip out stand for bench top use
- Rugged protective casing for field use and built-in electrode holder for safe storage
Typical applications include: Drinking water, cooling/boiler water, waste water, pool water, surface water, water treatment companies, industrial and governmental laboratories.
Delivery information: Supplied with batteries, pH electrode, temperature probe, pH buffer set 4,00 / 7,00, instruction manual, guarantee sheet and sturdy case.
The pH / Con set additionally includes a conductivity probe.
The pH / Oxi set additionally includes an oxygen sensor, electrolyte, and membrane heads.
The pH / Con / Oxi set additionally includes a conductivity probe, an oxygen sensor, electrolyte, and membrane heads.
Specifications
- Description:SensoDirect 150, pH / Oxi set, with pH electrode, oxygen sensor, temperature and conductivity probe
- Seq:03
- DO concentration resolution:0,1 % / 0,1 mg/L
- Conductivity resolution:0,1 µS (0 - 200,0 µS) / 0,001 mS (0,2 - 2,000 mS) / 0,01 mS (2 - 20,00 mS) / 0,1 mS (20 - 200,00 mS)
- DO accuracy:±0,4 mg/l / ± 0,7%
- Data logging capacity:16000 data sets
- Power supply:4×1,5 V AA battery or DC 9 V adapter
- mV resolution:1
- TDS accuracy:± 2 % F.S. + 1 digit
- Conductivity accuracy:± 2 % F.S. + 1 digit
- W×D×H:76×38×203 mm
- Temperature range:0...+60 °C
- mV range:±1999
- Conductivity reference temperature:0 - 60 °C
- pH range:0 - 14,00
- Temperature compensation:Automatic/manual (0...+100 °C)
- pH calibration:3-point calibration; pH 7, pH 4, and pH 10
- Temperature resolution:0,1 °C
- DO concentration:0 - 20,0 mg/L
- TDS resolution:0,1 / 1 / 10 / 100 ppm
- Temperature accuracy:± 0,8 °C
- Weight:0,39 kg (without protective covering)
- Cat. No.:LOVI724220
- pH accuracy:±0,02 + 2 digits
- pH resolution:0,01
- Data logging - log function:Automatic or manual
- Model:SensoDirect 150
- Conductivity range:0 - 200,0 µS / 0,2 - 2,000 mS / 2 - 20,00 mS / 20 - 200,00 mS
- Outputs:RS232
- TDS range:0 - 132 ppm / 132 - 1320 ppm / 1320 - 13200 ppm / 13200 - 132000 ppm
- mV accuracy:±0,5 % + 2 digits
- DO saturation:0 - 100,0 %
- Display:Large LCD with contrast adjustment
Accessories
Product Information
- Catalog #AvailabilityPrice
- Spare electrolyte for oxygen sensor
Product Information
- Catalog #AvailabilityPrice
- Power supply, AC220/230 V
Product Information
- Catalog #AvailabilityPrice
- Spare membrane for oxygen sensor
Product Information
- Catalog #AvailabilityPrice
- Case with foam
Product Information
- Catalog #AvailabilityPrice
- Temperature probe PT1000, with approx. 1,5 m cableTemperature probe PT1000, with approx. 1,5 m cableLOVI724420Log in for availability1 * 1 ST - SFr. 71.60
Product Information
- Catalog #AvailabilityPrice
- Oxygen sensor, with approx. 4 m cable
Product Information
- Catalog #AvailabilityPrice
- Conductivity probe (Con/TDS), with approx. 1,2 m cableConductivity probe (Con/TDS), with approx. 1,2 m cableLOVI724400Log in for availability1 * 1 ST - SFr. 117.00
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



