Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Multi-parameter meter (pH/conductivity/oxygen), handheld, SensoDirect 150 | Avantor

Multi-parameter meter (pH/conductivity/oxygen), handheld, SensoDirect 150

SensoDirect 150, pH / Con set, with pH electrode, oxygen sensor, temperature and conductivity probe
Katalognr. LOVI724210
Anbieter:   Lovibond
Multi-parameter meter (pH/conductivity/oxygen), handheld, SensoDirect 150
Katalognr. LOVI724210
Anbieter:  Lovibond
Multi-parameter meter (pH/conductivity/oxygen), handheld, SensoDirect 150
Multi-parameter meter (pH/conductivity/oxygen), handheld, SensoDirect 150

SensoDirect 150, pH / Con set, with pH electrode, oxygen sensor, temperature and conductivity probe

Preis
SFr. 1’130.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

The SensoDirect 150 is designed for multi purpose operation and measures pH/Redox, dissolved oxygen and conductivity/TDS.

  • Data logger function, results can be downloaded to a PC for further analysis
  • Automatic calibration + buffer recognition
  • Large LCD display, intuitive user interface, HOLD function
  • Flip out stand for bench top use
  • Rugged protective casing for field use and built-in electrode holder for safe storage

Typical applications include: Drinking water, cooling/boiler water, waste water, pool water, surface water, water treatment companies, industrial and governmental laboratories.

Lieferinformationen: Supplied with batteries, pH electrode, temperature probe, pH buffer set 4,00 / 7,00, instruction manual, guarantee sheet and sturdy case.
The pH / Con set additionally includes a conductivity probe.
The pH / Oxi set additionally includes an oxygen sensor, electrolyte, and membrane heads.
The pH / Con / Oxi set additionally includes a conductivity probe, an oxygen sensor, electrolyte, and membrane heads.

Technische Daten
  • Description:
    SensoDirect 150, pH / Con set, with pH electrode, oxygen sensor, temperature and conductivity probe
  • Seq:
    02
  • DO concentration resolution:
    0,1 % / 0,1 mg/L
  • Conductivity resolution:
    0,1 µS (0 - 200,0 µS) / 0,001 mS (0,2 - 2,000 mS) / 0,01 mS (2 - 20,00 mS) / 0,1 mS (20 - 200,00 mS)
  • DO accuracy:
    ±0,4 mg/l / ± 0,7%
  • Data logging capacity:
    16000 data sets
  • Power supply:
    4×1,5 V AA battery or DC 9 V adapter
  • mV resolution:
    1
  • TDS accuracy:
    ± 2 % F.S. + 1 digit
  • Conductivity accuracy:
    ± 2 % F.S. + 1 digit
  • W×D×H:
    76×38×203 mm
  • Temperature range:
    0...+60 °C
  • mV range:
    ±1999
  • Conductivity reference temperature:
    0 - 60 °C
  • pH range:
    0 - 14,00
  • Temperature compensation:
    Automatic/manual (0...+100 °C)
  • pH calibration:
    3-point calibration; pH 7, pH 4, and pH 10
  • Temperature resolution:
    0,1 °C
  • DO concentration:
    0 - 20,0 mg/L
  • TDS resolution:
    0,1 / 1 / 10 / 100 ppm
  • Temperature accuracy:
    ± 0,8 °C
  • Weight:
    0,39 kg (without protective covering)
  • Cat. No.:
    LOVI724210
  • pH accuracy:
    ±0,02 + 2 digits
  • pH resolution:
    0,01
  • Data logging - log function:
    Automatic or manual
  • Model:
    SensoDirect 150
  • Conductivity range:
    0 - 200,0 µS / 0,2 - 2,000 mS / 2 - 20,00 mS / 20 - 200,00 mS
  • Outputs:
    RS232
  • TDS range:
    0 - 132 ppm / 132 - 1320 ppm / 1320 - 13200 ppm / 13200 - 132000 ppm
  • mV accuracy:
    ±0,5 % + 2 digits
  • DO saturation:
    0 - 100,0 %
  • Display:
    Large LCD with contrast adjustment
Accessories

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • LOVI724470
    Spare electrolyte for oxygen sensor
    Einloggen für Verfügbarkeit
    1 * 50 mL - SFr. 30.30

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • LOVI724540
    Power supply, AC220/230 V
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 52.60

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • LOVI724460
    Spare membrane for oxygen sensor
    Einloggen für Verfügbarkeit
    1 * 2 ST - SFr. 49.60

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • LOVI725050
    Case with foam
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 85.60

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • LOVI724420
    Temperature probe PT1000, with approx. 1,5 m cable
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 71.60

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • LOVI724410
    Oxygen sensor, with approx. 4 m cable
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 284.00

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • LOVI724400
    Conductivity probe (Con/TDS), with approx. 1,2 m cable
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 117.00

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen..