Order Entry
Switzerland
Orders LinkContactUsLinkComponent
(2-Hydroxylpropyl)-β-cyclodextrine

(2-Hydroxylpropyl)-β-cyclodextrine

Glass bottle for solids, 100 g
Catalog # H31133.22
(2-Hydroxylpropyl)-β-cyclodextrine
Catalog # H31133.22
CAS Number:  128446-35-5
(2-Hydroxylpropyl)-β-cyclodextrine
(2-Hydroxylpropyl)-β-cyclodextrine

Glass bottle for solids, 100 g

CAS Number:  128446-35-5
Price
SFr. 749.00
Log in for availability
1 * 100 g
Product Details & Documents

CAS No.: 128446-35-5 Melting Point: 305° MDL No.: MFCD00069372

Synonyms:  Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
  • CAS Number:128446-35-5
  • Formula:C₆₃H₁₁₂O₄₂
  • MDL Number:MFCD16621721
  • Melting Pt:305 °C
  • Storage Temperature:Ambient
  • EINECS:420-920-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.