Orders LinkContactUsLinkComponent
Blanking nuts
fit port size ¼" NPT
Catalog # COWIPC702.025
Supplier:   COWIE
Blanking nuts
Catalog # COWIPC702.025
Supplier:  COWIE
Blanking nuts
Blanking nuts
fit port size ¼" NPT
Price
SFr. 10.30
Log in for availability
1 * 1 ST
Product Details & Documents
PTFE, white.
  • Chemically inert
  • Reduce contamination and simplify cleaning
  • Non-scratch
NPT blanking nuts/plugs are used to seal unused ports. To assemble simply screw the NPT blanking nut into a corresponding NPT port of the reactor lid.
Specifications
  • Description:
    fit port size ¼" NPT
  • Seq:
    01