Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Filter discs, DURAN®

Filter discs, DURAN®

5, 25 mm, 3,4 mm
Catalog # SCOT251525508
Supplier:   DWK Life Sciences
Filter discs, DURAN®
Catalog # SCOT251525508
Supplier:  DWK Life Sciences
Filter discs, DURAN®
Filter discs, DURAN®

5, 25 mm, 3,4 mm

Price
SFr. 294.00
Log in for availability
1 * 10 ST
Product Details & Documents

Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.

  • Available in intermediate sizes, alternative dimensions, alternate porosities and bi-planar design

Made from clear DURAN® borosilicate glass 3,3.

Specifications
  • Ø:
    25 mm
  • Porosity:
    5
  • Seq:
    89
  • Thickness:
    3,4 mm
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..