ContactUsLinkComponent
Full option autoclave, fully automatic, microprocessor-controlled vertical laboratory autoclave with Cooling Sytem, Vacuum system,Printer and EU-plug, Chamberdimension (xL) 380 x 690 mm Volume: 85 liters 1 * 1 items
Catalog # TUTT3870ELV-FO-EU
Supplier:  TUTTNAUER
Full option autoclave, fully automatic, microprocessor-controlled vertical laboratory autoclave with Cooling Sytem, Vacuum system,Printer and EU-plug, Chamberdimension (xL) 380 x 690 mm Volume: 85 liters 1 * 1 items
Catalog # TUTT3870ELV-FO-EU
Supplier:  TUTTNAUER
Frequently Bought Together