Glass beads
Glass beads, solid, 500 g pack, 4 mm
Made of soda lime glass.
- Solid
- Polished
Glass beads are used for mixing or milling. The smaller the beads and the higher their quantity the more intense is the mixing or milling.
: In poly bag of 500 g or 1000 g each.
- Ø:4 mm
- Description:Glass beads, solid, 500 g pack
- Seq:04



