Labels, pressure-sensitive
Labels, Polyolefin, White, 13×44 mm
Catalog # 817-0084
Supplier: Thermo Fisher Scientific
Labels, pressure-sensitive
Catalog # 817-0084
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These Nalgene™ polyolefin pressure-sensitive labels are used with photocopiers, laser printers or inkjet printers. Accepts a variety of inks including copier, marker, ballpoint, gels, and technical markers (pretest specific inks for suitability prior to use), does not accept pencil.
- Rated to −29 °C
- Resistant to ultraviolet light, mildew, chemicals and dirt and won't discolor, crack or tear
- Excellent for labeling microscopic slides - adhesive backing sticks well to any plastic, glass or metal surface
- Chemically compatible with most lab reagents although solvents may dissolve adhesive
- Autoclavable
Formatted for easy setup in word processing programs; instructions included in each pack.
Specifications
- Colour:White
- Description:Labels
- Dimensions:13×44 mm
- Material:Polyolefin
- Cat. No.:817-0084
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact



