Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Compact automatic burettes, Schellbach stripe, SILBERBRAND

Compact automatic burettes, Schellbach stripe, SILBERBRAND

50 ml, 0,1 ml, 730 mm, 0,10 ± ml
Catalog # BRND23811
Supplier:   Brand
Compact automatic burettes, Schellbach stripe, SILBERBRAND
Catalog # BRND23811
Supplier:  Brand
Compact automatic burettes, Schellbach stripe, SILBERBRAND
Compact automatic burettes, Schellbach stripe, SILBERBRAND

50 ml, 0,1 ml, 730 mm, 0,10 ± ml

Price
SFr. 151.00
Log in for availability
1 * 1 ST
Product Details & Documents

AR® glass, class B.

  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, reduced distance between subdivision marks
  • Detachable PTFE stopcock

Error limits according to class B, DIN EN ISO 385.

Ordering information: Please order supports and burette clamps separately.

Delivery information: Supplied as burette tube, stopcock with precision tip, filling tube (PVC) and 1000 ml PE bottle with base.

Specifications
  • Capacity:
    50 ml
  • Length:
    730 mm
  • Division:
    0,1 ml
  • Tolerance:
    0,10 ± ml
Accessories

Product Information

  • Catalog #
    Capacity
    Availability
    Price
  • Reservoir bottle, LDPE, GL 28
    Reservoir bottle, LDPE, GL 28
    BRND129060
    1000 ml
    Log in for availability
    1 * 1 ST - SFr. 9.00

Product Information

  • Catalog #
    Capacity
    Availability
    Price
  • Bases for bottles
    Bases for bottles
    BRND23728
    1000 ml
    Log in for availability
    1 * 1 ST - SFr. 9.70

Product Information

  • Catalog #
    Capacity
    Availability
    Price
  • Bases for bottles
    Bases for bottles
    BRND23725
    500 ml
    Log in for availability
    1 * 1 ST - SFr. 8.10

Product Information

  • Catalog #
    Availability
    Price
  • Spare burette stopcock, PTFE
    Spare burette stopcock, PTFE
    BRND11805
    Log in for availability
    1 * 1 ST - SFr. 56.90

Product Information

  • Catalog #
    Availability
    Price
  • Spare burette tip, clear borosilicate 3.3 glass, for 10 ml compact burettes and compact automatic burettes
    Spare burette tip, clear borosilicate 3.3 glass, for 10 ml compact burettes and compact automatic burettes
    BRND11510
    Log in for availability
    1 * 1 ST - SFr. 7.80

Product Information

  • Catalog #
    Availability
    Price
  • Spare burette tip, clear borosilicate 3.3 glass, for 25 ml compact burettes and compact automatic burettes
    Spare burette tip, clear borosilicate 3.3 glass, for 25 ml compact burettes and compact automatic burettes
    BRND11511
    Log in for availability
    1 * 1 ST - SFr. 7.80

Product Information

  • Catalog #
    Availability
    Price
  • Spare burette tip, clear borosilicate 3.3 glass, for 50 ml compact burettes and compact automatic burettes
    Spare burette tip, clear borosilicate 3.3 glass, for 50 ml compact burettes and compact automatic burettes
    BRND11512
    Log in for availability
    1 * 1 ST - SFr. 7.80

Product Information

  • Catalog #
    Capacity
    Availability
    Price
  • Reservoir bottle, LDPE, GL 25
    Reservoir bottle, LDPE, GL 25
    BRND129055
    500 ml
    Log in for availability
    1 * 1 ST - SFr. 5.10

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.