Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Buffer solutions, colour coded, Fluka™

7,00, Green, 20 °C, Plastic bottle, 6×0,5 L, 3.000 mL
Catalog # 33666-6X500ML
Supplier:   Honeywell Chemicals
Buffer solutions, colour coded, Fluka™
Catalog # 33666-6X500ML
CAS Number:  
Buffer solutions, colour coded, Fluka™

7,00, Green, 20 °C, Plastic bottle, 6×0,5 L, 3.000 mL

CAS Number:  
Price
316.24 SFr.
Log in for availability
1 * 3.000 mL
Product Details & Documents

These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.

 

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    3.000 mL
  • Reference temperature:
    20 °C
  • Colour:
    Green
  • Pk Info:
    6×0,5 L
  • pH value:
    7,00

Specifications