Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer solutions, colour coded, Fluka™

Buffer solutions, colour coded, Fluka™

4,00, Red, 20 °C, Plastic bottle, 6×0,5 L, 3.000 mL
Catalog # 33665-6X500ML
Supplier:   Honeywell Chemicals
Buffer solutions, colour coded, Fluka™
Catalog # 33665-6X500ML
CAS Number:  
Buffer solutions, colour coded, Fluka™

4,00, Red, 20 °C, Plastic bottle, 6×0,5 L, 3.000 mL

CAS Number:  
Price
SFr. 327.95
Log in for availability
1 * 3.000 mL
Product Details & Documents

These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    3.000 mL
  • Reference temperature:
    20 °C
  • Colour:
    Red
  • Pk Info:
    6×0,5 L
  • pH value:
    4,00

Specifications