Order Entry
Switzerland
Orders LinkContactUsLinkComponent
VWR®, Test Tubes

VWR®, Test Tubes

3,5 ml, 12,0 mm, PS, 11,8 mm
Catalog # 212-1837
Supplier:   VWR Collection
VWR®, Test Tubes
Catalog # 212-1837
Supplier:  VWR Collection
VWR®, Test Tubes
VWR®, Test Tubes

3,5 ml, 12,0 mm, PS, 11,8 mm

Price
SFr. 158.00
Log in for availability
1 * 5.000 ST
Product Details & Documents

Polystyrene (PS), transparent.

  • Can be centrifuged up to 3000×g
  • Can be delivered sterile with red plug or non sterile without plug
  • Additional plugs and other colours available upon request

Universal tubes with round bottom for a wide range of applications.

Specifications
  • Ext.Ø:
    12,0 mm
  • Ø int.:
    11,8 mm
  • Capacity:
    3,5 ml
  • Material:
    PS
  • Seq:
    01
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.