Orders LinkContactUsLinkComponent
Liquid nitrogen level control, auto-fill system
Liquid nitrogen level control, auto-fill system
Magnetic valve for liquid nitrogen, 24 V
Catalog # KGWA1303
Supplier:   KGW
Liquid nitrogen level control, auto-fill system
Catalog # KGWA1303
Supplier:  KGW
Liquid nitrogen level control, auto-fill system
Liquid nitrogen level control, auto-fill system
Magnetic valve for liquid nitrogen, 24 V
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Level control for liquid nitrogen type is manual or automatic refill. It is used for level control of liquid nitrogen (LN₂) in unsealed Dewar flasks.
  • Parting of cable safety device for sensor
  • Optical indication of the actual state
Applicable for acoustic and optical error indication, the controller opens whole refilling the magnetic valve at the reservoir tank.
Delivery information: Supplied with two temperature sensors PT 100 (minimum, maximum sensors), socket for LN₂-magnetic valve (24 V), function control for sensor.
Specifications
  • Description:
    Magnetic valve for liquid nitrogen, 24 V
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..