Orders LinkContactUsLinkComponent
Adapters, delivery, with vent, QUICKFIT®
Adapters, delivery, with vent, QUICKFIT®
19/26 NS, 19/26 NS
Catalog # 271-0009
Supplier:   SciLabware
Adapters, delivery, with vent, QUICKFIT®
Catalog # 271-0009
Supplier:  SciLabware
Adapters, delivery, with vent, QUICKFIT®
Adapters, delivery, with vent, QUICKFIT®
19/26 NS, 19/26 NS
Price
SFr. 62.50
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass, bend.
  • Avoids the problems of 'closed systems'
With a side arm for venting purposes, not for connection to a vacuum source. For adapters with vacuum connection, see 201-8653.
Specifications
  • Cone size:
    19/26 NS
  • Socket size:
    19/26 NS
  • Seq:
    02