Orders LinkContactUsLinkComponent
Forceps, precision
Forceps, precision
Precision forceps, extra sharp, straight (tower tip), Straight, 105 mm
Catalog # BOCH1930
Supplier:   BOCHEM
Forceps, precision
Catalog # BOCH1930
Supplier:  BOCHEM
Forceps, precision
Forceps, precision
Precision forceps, extra sharp, straight (tower tip), Straight, 105 mm
Price
SFr. 76.60
Log in for availability
1 * 1 ST
Product Details & Documents
These non serrated forceps are made of durable titanium.
  • High corrosion and thermal resistance
  • Lightweight, high strength
Titanium is resistant to diluted sulphuric acid, hydrochloric acid, solutions containing chloride, cold nitric acid as well as most organic acids and alkaline solutions, such as sodium hydroxide.
Specifications
  • Type:
    Straight
  • Description:
    Precision forceps, extra sharp, straight (tower tip)
  • Length:
    105 mm
  • Seq:
    01