Orders LinkContactUsLinkComponent
Support clips
Support clips
Support clip with fitting, O. Ø 58 - 61 mm
Catalog # BOCH594
Supplier:   BOCHEM
Support clips
Catalog # BOCH594
Supplier:  BOCHEM
Support clips
Support clips
Support clip with fitting, O. Ø 58 - 61 mm
Price
SFr. 104.00
Log in for availability
1 * 1 ST
Product Details & Documents
These support clips are coated with ceramic inside.
  • For tubes and cylindrical vessels
  • With fitting
Ordering information: The shafts (rods) need to be ordered separately.
Specifications
  • Description:
    Support clip with fitting, O. Ø 58 - 61 mm