Support clips
Support clip with fitting, O. Ø 40 - 44 mm
Catalog # BOCH592
Supplier: BOCHEM
Jump to:
Product Details & Documents
These support clips are coated with ceramic inside.
- For tubes and cylindrical vessels
- With fitting
Ordering information: The shafts (rods) need to be ordered separately.
Specifications
- Description:Support clip with fitting, O. Ø 40 - 44 mm
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.