Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cryoboxes

Cryoboxes

5×5, Tubes 11,0 mm Ø, White, 68×68×30 mm
Catalog # RATI5160036
Supplier:   RATIOLAB
Cryoboxes
Catalog # RATI5160036
Supplier:  RATIOLAB
Cryoboxes
Cryoboxes

5×5, Tubes 11,0 mm Ø, White, 68×68×30 mm

Price
SFr. 24.70
Log in for availability
1 * 10 ST
Product Details & Documents

Cryoboxes, cardboard, for storage of cryovials at low temperatures.

  • Waterproof coating
  • Dividers must to be ordered separately
  • Available in different heights and five colours
Specifications
  • Compartments:
    5×5
  • For:
    Tubes 11,0 mm Ø
  • W×D×H ext.:
    68×68×30 mm
  • Colour:
    White
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.