ContactUsLinkComponent
Pipettenspitze 0.5-10 µl, MµltiFlex E flach 0,2 mm, Rack/200 Stk SORENSON (200 Stk) 200 Stk 1 * 200 ST
Catalog # SORE17330
Pipettenspitze 0.5-10 µl, MµltiFlex E flach 0,2 mm, Rack/200 Stk SORENSON (200 Stk) 200 Stk 1 * 200 ST
Catalog # SORE17330
Frequently Bought Together