Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
QuickRack, 1 - 200 µl, Yellow
Catalog # SORE30460
Supplier:   Sorenson Bioscience
Pipette tips
Catalog # SORE30460
Pipette tips
QuickRack, 1 - 200 µl, Yellow
Price
SFr. 80.90
Log in for availability
1 * 960 ST
Product Details & Documents
These universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
Specifications
  • Volume:
    1 - 200 µl
  • Seq:
    03
  • Tip style:
    QuickRack
  • Tip colour:
    Yellow