Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Gas sampling balloons

Gas sampling balloons

Gas sampling balloon, 160 mm, 2,0 L
Catalog # KART1232
Supplier:   KARTELL
Gas sampling balloons
Catalog # KART1232
Supplier:  KARTELL
Gas sampling balloons
Gas sampling balloons

Gas sampling balloon, 160 mm, 2,0 L

Price
SFr. 43.80
Log in for availability
1 * 1 pezzi
Product Details & Documents

Gas collection bags made from latex.

Designed for general use in laboratories.

Ordering information: Available in different volumes and diameters.

Documents

Specifications
  • Ø:
    160 mm
  • Volume:
    2,0 L
  • Description:
    Gas sampling balloon
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.