Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Ammonia buffer solution pH10 (20 °C) for complexometry, Fluka™

Ammonia buffer, 10, Poly bottle, 6 L
Catalog # 33582-6X1L
Supplier:   Honeywell Chemicals
Ammonia buffer solution pH10 (20 °C) for complexometry, Fluka™
Catalog # 33582-6X1L
CAS Number:  
Ammonia buffer solution pH10 (20 °C) for complexometry, Fluka™

Ammonia buffer, 10, Poly bottle, 6 L

CAS Number:  
Price
SFr. 473.52
Log in for availability
1 * 6 L
Product Details & Documents

 

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    6 L
  • Description:
    Ammonia buffer
  • pH value:
    10

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.