Assay Kit SIRT1 Direct flour Screening 96well 1 * 1 KIT
Catalog # CAYM10010401-96
Anbieter: Cayman Chemical
Assay Kit SIRT1 Direct flour Screening 96well 1 * 1 KIT
Catalog # CAYM10010401-96
Anbieter: Cayman Chemical
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
Anbieter: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Anbieter: VWR Collection
VWR® Facial Tissues
Anbieter: VWR Collection
VWR®, Nitrile Gloves
Anbieter: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Anbieter: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Sealing film for manual application, Parafilm® M
Anbieter: HEATHROW
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Anbieter: VWR Collection
VWR®, Transfer Pipettes, High Performance
Anbieter: VWR Collection