Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Storage Boxes with Lid

Storage Boxes with Lid

1,5 l, Transparent, 200×100×85 mm, PS
Catalog # SEMA0881
Supplier:   SEMADENI
Storage Boxes with Lid
Catalog # SEMA0881
Supplier:  SEMADENI
Storage Boxes with Lid
Storage Boxes with Lid

1,5 l, Transparent, 200×100×85 mm, PS

Price
SFr. 10.00
Log in for availability
1 * 1 ST
Product Details & Documents

These storage boxes with transparent lids are either made of polystyrene (PS) or styrene-acrylonitrile resin (SAN).

Specifications
  • Capacity:
    1,5 l
  • Colour:
    Transparent
  • Material:
    PS
  • Seq:
    02
  • W×D×H:
    200×100×85 mm