Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Sample concentrators, Vivaspin®

Sample concentrators, Vivaspin®

50 kD
Catalog # 28-9322-36
Supplier:   Cytiva
Sample concentrators, Vivaspin®
Catalog # 28-9322-36
Supplier:  Cytiva
Sample concentrators, Vivaspin®
Sample concentrators, Vivaspin®

50 kD

Price
SFr. 162.00
Log in for availability
1 * 25 ST
Product Details & Documents

Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration.

  • One-step sample concentration in a single tube for minimal sample handling and reduced sample loss
  • Patented dead-stop technology, which ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery
  • Vertical PES membrane minimises membrane blockage and tolerates high flow rates
  • Compatible pH range is from pH 1 to 9

Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. The entire process is performed in a single tube with an upper compartment containing sample and lower compartment separated by a semipermeable membrane with a molecular weight cutoff (MWCO) selected by the user. Centrifugation is applied to force solvent through the membrane, leaving a more concentrated sample in the upper chamber.

Vivaspin® sample concentrators cater for sample volumes from 100 μl to 20 ml, with a range of molecular weight cutoff values from Mr 3000 to 100000. For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.

Specifications
  • MWCO:
    50 kD
  • Seq:
    05
  • Filtration area:
    0,5 cm²
  • Cat. No.:
    28-9322-36
  • Type:
    Vivaspin® 500
  • Dead volume:
    <5 µl
  • Ø×H:
    11×50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.