Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Titration beakers

100 ml, Titration beaker, PP, 20 ml
  661-0120
Titration beakers
  661-0120
Titration beakers

100 ml, Titration beaker, PP, 20 ml

 
329.00 SFr.
   
1 * 480 ST
 

Disposable titration beakers made from Polypropylene.

  • Flexible, not breakable like traditional glass lab beakers
  • Disposable after use
  • Convenient 20 ml graduations make quick measurements easier
  • Good chemical resistance for the majority of common lab applications
 
  • Graduation Interval:
    20 ml
  • Capacity:
    100 ml
  • Description:
    Titration beaker, PP
  • Seq:
    10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.