Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Accessories

Accessories

Seal for switch button, Ø 21 mm, G 7855, G 7856, G 7857, G 7858, G 7859...
Catalog # 141-0841
Accessories
Catalog # 141-0841
Supplier:  
Accessories
Accessories

Seal for switch button, Ø 21 mm, G 7855, G 7856, G 7857, G 7858, G 7859...

Discontinued
Product Details & Documents
  • For lower and upper baskets
  • With 7 x 7 mm perforations
  • Stainless steel
Specifications
  • Description:
    Seal for switch button, Ø 21 mm
  • Seq:
    30
  • For:
    G 7855, G 7856, G 7857, G 7858, G 7859...
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.